Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mier3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172593.3 | NP_766181.2 | 3, 4 | 356 | ||
GenBank mRNA | AK030747.1 | - | 3, 4 | 284 | ||
AK083929.1 | - | 2, 3 | 195 | |||
AK147427.1 | - | 3, 4 | 357 | |||
BC103780.1 | - | 2, 3 | 186 | 30108551 | ||
BC138127.1 | - | 2, 3 | 176 | 8861146 |