Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172497.3 | NP_766085.2 | 1 | 179 | ||
GenBank mRNA | AK029550.1 | - | 1 | 180 | ||
AK167235.1 | - | 1 | 163 | |||
BC132103.1 | - | 1 | 170 | 40130380 | ||
BC137880.1 | - | 1 | 170 | 8860899 |