Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF2AK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134335.1 | NP_001127807.1 | 2, 3 | 421 | ||
NM_014413.3 | NP_055228.2 | 2, 3 | 421 | |||
GenBank mRNA | AB037790.1 | - | 2, 3 | 346 | ||
AF100784.1 | - | 2, 3 | 310 | |||
AF147094.1 | - | 2, 3 | 356 | |||
AF183414.1 | - | 2, 3 | 308 | |||
AF255050.1 | - | 2, 3 | 366 | |||
AK075192.1 | - | 2, 3 | 349 | |||
AK290327.1 | - | 2, 3 | 354 | |||
AL136563.1 | - | 2, 3 | 326 | |||
AL834494.1 | - | 2, 3 | 326 | |||
BC006524.2 | - | 2, 3 | 353 | 2989989 | ||
DC341983.1 | - | 2, 3 | 421 |