Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfsd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114662.1 | NP_001108134.1 | 2 | 573 | ||
NM_172510.4 | NP_766098.2 | 2 | 573 | |||
GenBank mRNA | AK038882.1 | - | 2 | 466 | ||
AK044667.1 | - | 2 | 446 | |||
AK085484.1 | - | 2 | 445 | |||
AK162495.1 | - | 2 | 477 | |||
BC058790.1 | - | 2 | 422 | 6395070 |