Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272070.1 | NP_001258999.1 | 2 | 274 | ||
NM_172511.4 | NP_766099.3 | 2 | 274 | |||
GenBank mRNA | AK046757.1 | - | 1 | 2372 | ||
AK052670.1 | - | 2 | 223 | |||
AK084998.1 | - | 2 | 222 | |||
BC058347.1 | - | 2 | 223 | 6820515 | ||
BY173258.1 | - | 2 | 226 | |||
BY281009.1 | - | 2 | 275 |