Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gemin5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166669.1 | NP_001160141.1 | 3 | 460 | ||
NM_001166670.1 | NP_001160142.1 | 3 | 460 | |||
NM_172558.3 | NP_766146.2 | 3 | 460 | |||
GenBank mRNA | AK049216.1 | - | 3 | 395 | ||
AK143536.1 | - | 3 | 409 | |||
BC137802.1 | - | 3 | 403 | 8860821 | ||
BC144921.1 | - | 3 | 398 | |||
BC144922.1 | - | 3 | 403 | 9053470 | ||
BY742908.1 | - | 3 | 461 |