Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205081.1 | NP_001192010.1 | 4 | 1099 | ||
NM_172570.5 | NP_766158.3 | 4 | 1099 | |||
GenBank mRNA | AK031572.1 | - | 4 | 1074 | ||
AK075838.1 | - | 4 | 528 | |||
BC023393.1 | - | 2 | 274 | |||
BC024468.1 | - | 2 | 143 | |||
BC043714.1 | - | 3 | 411 | |||
BC055038.1 | - | 2 | 399 | |||
BC064728.1 | - | 4 | 493 | |||
BC080731.1 | - | 4 | 495 | |||
BC100317.1 | - | 4 | 1083 | |||
BC132148.1 | - | 4 | 880 | 40130425 | ||
BC137953.1 | - | 4 | 880 | 8860972 | ||
BC145059.1 | - | 4 | 878 | 9053607 |