Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lhfpl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172589.2 | NP_766177.1 | 3 | 750 | ||
GenBank mRNA | AK030103.1 | - | 2 | 908 | ||
AK031635.1 | - | 3 | 748 | |||
AK052969.1 | - | 1 | 612 | |||
AK087506.1 | - | 3 | 751 | |||
BC040389.1 | - | 1 | 534 | 3484974 |