Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcho2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172591.3 | NP_766179.2 | 4 | 646 | ||
GenBank mRNA | AK031041.1 | - | 4 | 649 | ||
AK142282.1 | - | 4 | 332 | |||
AK146566.1 | - | 4 | 380 | |||
AK164135.1 | - | 4 | 392 | |||
BC019933.1 | - | 5 | 382 | |||
BC047122.1 | - | 5 | 382 | |||
BC052456.1 | - | 4 | 343 | 6830607 | ||
BC053718.1 | - | 4 | 362 |