Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3bgrl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172507.5 | NP_766095.1 | 4 | 3140 | ||
GenBank mRNA | AK047567.1 | - | 1029 | |||
AK049293.1 | - | 1 | 2174 | |||
AK077373.1 | - | 1 | 1433 | |||
BC038473.1 | - | 4 | 3177 | 5008777 |