Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172509.3 | NP_766097.1 | 4 | 2958 | ||
GenBank mRNA | AK029942.1 | - | 4 | 2935 | ||
AK034583.1 | - | 4 | 2934 | |||
AK083583.1 | - | 4 | 2959 | |||
AK140606.1 | - | 4 | 2933 | |||
AK162568.1 | - | 4 | 2987 |