Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146048.1 | NP_001139520.1 | 8, 9 | 1017 | ||
NM_172528.3 | NP_766116.3 | 8, 9 | 665 | |||
GenBank mRNA | AK051043.1 | - | 8, 9 | 1016 | ||
AK082721.1 | - | 8, 9 | 666 | |||
AK147075.1 | - | 8, 9 | 1017 | |||
AK220166.1 | - | 8, 9 | 1035 | |||
BC046591.1 | - | 8, 9 | 1004 | |||
BC087542.1 | - | 8, 9 | 864 | 4934980 |