Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172532.3 | NP_766120.1 | 9, 10 | 1467 | ||
GenBank mRNA | AK052703.1 | - | 9, 10 | 1468 | ||
AK144030.1 | - | 9, 10 | 1451 | |||
AK159485.1 | - | 9, 10 | 1447 | |||
AK166313.1 | - | 9, 10 | 1447 | |||
BC138703.1 | - | 9, 10 | 1430 | 8861725 |