Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papolg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172555.2 | NP_766143.2 | 16 | 1671 | ||
GenBank mRNA | AK035810.1 | - | 16 | 1654 | ||
BC019635.1 | - | 1 | 24 | |||
BC037439.1 | - | 1 | 56 | |||
BC049163.1 | - | 16 | 1617 | |||
BC059271.1 | - | 16 | 1665 | 2811338 | ||
DQ103505.1 | - | 16 | 1477 |