Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntrob | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172560.3 | NP_766148.2 | 14 | 3017 | ||
GenBank mRNA | AK036710.1 | - | 13 | 2837 | ||
AK147774.1 | - | 14 | 2907 | |||
BC058512.1 | - | 14 | 1850 | 6817014 |