Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcho2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172591.3 | NP_766179.2 | 10 | 1203 | ||
GenBank mRNA | AK031041.1 | - | 10 | 1206 | ||
AK142282.1 | - | 10 | 889 | |||
AK146566.1 | - | 10 | 937 | |||
AK164135.1 | - | 10 | 949 | |||
BC052456.1 | - | 10 | 900 | 6830607 | ||
BC053718.1 | - | 10 | 919 |