Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tomm22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172609.3 | NP_766197.2 | 4 | 702 | ||
GenBank mRNA | AK008133.1 | - | 4 | 705 | ||
AK009868.1 | - | 4 | 702 | |||
AK013471.1 | - | 4 | 691 | |||
AK049442.1 | - | 690 | ||||
AK150360.1 | - | 4 | 690 | |||
AK150787.1 | - | 4 | 699 | |||
AK152843.1 | - | 4 | 690 | |||
AK167803.1 | - | 4 | 694 | |||
AK210532.1 | - | 1 | 42 | |||
BC056920.1 | - | 4 | 684 | 6819748 |