Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp13a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164612.1 | NP_001158084.1 | 11 | 1375 | ||
NM_001164613.1 | NP_001158085.1 | 12 | 1428 | |||
NM_172613.4 | NP_766201.3 | 11 | 1375 | |||
GenBank mRNA | AK028776.1 | - | 12 | 1430 | ||
AK029303.1 | - | 10 | 1119 | |||
BC048410.1 | - | 11 | 1375 | 6486835 |