Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMELX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142.2 | NP_001133.1 | 6 | 697 | ||
NM_182680.1 | NP_872621.1 | 7 | 739 | |||
NM_182681.1 | NP_872622.1 | 5 | 649 | |||
GenBank mRNA | AF436849.1 | - | 7 | 739 | ||
M86932.1 | - | 6 | 697 |