Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK032112.1 | - | 1, 2 | 387 | ||
AK042555.1 | - | 2, 3 | 662 | |||
BC055682.1 | - | 3, 4 | 660 | 6412317 |