Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172662.3 | NP_766250.2 | 3 | 216 | ||
GenBank mRNA | AK028468.1 | - | 3 | 139 | ||
AK038057.1 | - | 2 | 87 | |||
AK054272.1 | - | 3 | 217 | |||
AK163943.1 | - | 3 | 174 |