Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172663.4 | NP_766251.3 | 1 | 397 | ||
GenBank mRNA | AK031025.1 | - | 1 | 370 | ||
AK047947.1 | - | 1 | 65 | |||
BB620197.1 | - | 1 | 359 |