Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gyltl1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290774.1 | NP_001277703.1 | 3, 4 | 338 | ||
GenBank mRNA | AK077630.1 | - | 3, 4 | 292 |