Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANPEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001150.2 | NP_001141.2 | 2 | 816 | ||
XM_005254892.4 | XP_005254949.1 | 2 | 824 | |||
XM_011521473.1 | XP_011519775.1 | 2 | 967 | |||
GenBank mRNA | AB209918.1 | - | 2 | 812 | ||
BC058928.1 | - | 1 | 617 | 6020902 | ||
M22324.1 | - | 1 | 644 | |||
X13276.1 | - | 1 | 548 |