Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oscp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172701.2 | NP_766289.2 | 2 | 270 | ||
GenBank mRNA | AB154826.1 | - | 2 | 223 | ||
AB286960.1 | - | 3 | 271 | |||
AK077467.1 | - | 2 | 236 | |||
AK077909.1 | - | 2 | 191 | |||
BC045150.1 | - | 2 | 216 | 6309923 |