Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANXA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001153.4 | NP_001144.1 | 4, 5 | 447 | ||
NM_001320698.1 | NP_001307627.1 | 6, 7 | 1729 | |||
NM_001320700.1 | NP_001307629.1 | 4, 5 | 447 | |||
NM_001320702.1 | NP_001307631.1 | 4, 5 | 482 | |||
XM_017003943.1 | XP_016859432.1 | 7, 8 | 1591 | |||
XM_017003944.1 | XP_016859433.1 | 7, 8 | 1557 | |||
GenBank mRNA | AB209457.1 | - | 4, 5 | 326 | ||
AK058017.1 | - | 4, 5 | 287 | |||
AK293177.1 | - | 4, 5 | 354 | |||
AK293396.1 | - | 4, 5 | 285 | |||
AK293406.1 | - | 4, 5 | 235 | |||
AK303960.1 | - | 4, 5 | 289 | |||
AK315511.1 | - | 4, 5 | 287 | |||
AY453398.1 | - | 3, 4 | 231 | |||
BC000182.1 | - | 4, 5 | 329 | 3352245 | ||
BC011659.1 | - | 4, 5 | 307 | 4301675 | ||
BC063672.1 | - | 4, 5 | 447 | 3908264 | ||
BE886769.1 | - | 426 | ||||
CR407681.1 | - | 2, 3 | 199 | |||
D78152.1 | - | 3, 4 | 261 | |||
FJ224287.1 | - | 4, 5 | 272 | |||
M19383.1 | - | 4, 5 | 272 | |||
M82809.1 | - | 4, 5 | 272 |