Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gramd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205353.1 | NP_001192282.1 | 2 | 318 | ||
NM_172611.4 | NP_766199.1 | 2 | 220 | |||
GenBank mRNA | AK036832.1 | - | 2 | 209 | ||
AK142266.1 | - | 2 | 173 | |||
BC085148.1 | - | 2 | 276 |