Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Piezo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039485.4 | NP_001034574.4 | 45, 46 | 7989 | ||
GenBank mRNA | AK031235.1 | - | 5, 6 | 927 | ||
AK031422.1 | - | 4, 5 | 648 | |||
BC026614.1 | - | 2, 3 | 377 | |||
BC075677.1 | - | 6, 7 | 1080 | |||
BC147606.1 | - | 4, 5 | 542 | 9056596 | ||
BC172109.1 | - | 4, 5 | 542 | |||
HQ215521.1 | - | 45, 46 | 7069 |