Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177627.1 | NP_001171098.1 | 5 | 824 | ||
NM_172659.2 | NP_766247.2 | 5 | 824 | |||
GenBank mRNA | AK079650.1 | - | 5 | 768 | ||
AK089246.1 | - | 5 | 816 | |||
AK137441.1 | - | 5 | 790 | |||
AK150104.1 | - | 5 | 777 | |||
AK163445.1 | - | 5 | 787 | |||
AK166312.1 | - | 5 | 782 | |||
AK167904.1 | - | 5 | 786 | |||
AK172411.1 | - | 5 | 768 | |||
BC025498.1 | - | 4 | 620 | |||
BC058210.1 | - | 5 | 748 | |||
BC141168.1 | - | 5 | 755 | 9055780 | ||
BC145480.1 | - | 5 | 755 |