Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172676.2 | NP_766264.2 | 5 | 1769 | ||
GenBank mRNA | AK053048.1 | - | 5 | 1722 | ||
AK053884.1 | - | 3 | 1350 | |||
AK164204.1 | - | 5 | 1809 | |||
AK171151.1 | - | 5 | 1775 | |||
AK212985.1 | - | 1 | 17 | |||
BC052921.1 | - | 5 | 1769 | 6311715 |