Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172685.3 | NP_766273.1 | 1, 2 | 244 | ||
GenBank mRNA | AK032806.1 | - | 1, 2 | 245 | ||
AK146034.1 | - | 1, 2 | 243 | |||
AK167778.1 | - | 1, 2 | 235 | |||
AK168090.1 | - | 1, 2 | 235 | |||
AK170598.1 | - | 1, 2 | 236 | |||
AK187917.1 | - | 13 | ||||
BC055369.1 | - | 1, 2 | 233 | 30055307 |