Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gba2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172692.3 | NP_766280.2 | 1 | 791 | ||
GenBank mRNA | AK089192.1 | - | 1 | 477 | ||
AK158174.1 | - | 1 | 791 | |||
AK173213.1 | - | 1 | 777 | |||
BC031397.1 | - | 1 | 464 | |||
BC034105.1 | - | 1 | 16 | |||
BC056935.1 | - | 1 | 314 | 5703478 |