Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txndc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172597.2 | NP_766185.1 | 4, 5 | 562 | ||
GenBank mRNA | AK017582.1 | - | 4, 5 | 457 | ||
AK034025.1 | - | 4, 5 | 551 | |||
AK039945.1 | - | 4, 5 | 429 | |||
AK046466.1 | - | 4, 5 | 563 | |||
AK173153.1 | - | 1059 | ||||
AK215424.1 | - | 2, 3 | 142 | |||
BC052488.1 | - | 4, 5 | 438 | 6438726 |