Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: March6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172606.2 | NP_766194.2 | 26 | 4949 | ||
GenBank mRNA | AK089827.1 | - | 20 | 2300 | ||
AK129169.1 | - | 14 | 3692 | |||
BC027068.1 | - | 1 | 1286 | |||
BC031721.1 | - | 2 | 2231 | |||
BC039776.1 | - | 1 | 1574 | |||
BC048816.1 | - | 13 | 3589 | |||
BC059190.1 | - | 12 | 3477 |