Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gramd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205353.1 | NP_001192282.1 | 19 | 4054 | ||
NM_172611.4 | NP_766199.1 | 19 | 3861 | |||
GenBank mRNA | AK036832.1 | - | 19 | 3851 | ||
AK129209.1 | - | 15 | 5831 | |||
AK142266.1 | - | 18 | 4610 | |||
BC021523.1 | - | 15 | 3236 | |||
BC085148.1 | - | 19 | 4012 | |||
BY596457.1 | - | 1 | 169 |