Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp13a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164612.1 | NP_001158084.1 | 30 | 3769 | ||
NM_172613.4 | NP_766201.3 | 30 | 3712 | |||
GenBank mRNA | AK029303.1 | - | 29 | 3513 | ||
BC038696.1 | - | 4 | 621 | |||
BC048410.1 | - | 30 | 3712 | 6486835 |