Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam214b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253353.1 | NP_001240282.1 | 9 | 2239 | ||
NM_001253354.1 | NP_001240283.1 | 10 | 2016 | |||
NM_172691.3 | NP_766279.1 | 9 | 2020 | |||
GenBank mRNA | AK045811.1 | - | 9 | 2004 | ||
AK083914.1 | - | 2 | 2383 | |||
AK149610.1 | - | 9 | 2097 | |||
AK154748.1 | - | 8 | 2355 | |||
AK161380.1 | - | 9 | 1947 | |||
AK173197.1 | - | 6 | 765 | |||
BC022749.1 | - | 3 | 432 | |||
BC044832.1 | - | 8 | 2177 | |||
BC046771.1 | - | 10 | 2006 | 5703628 | ||
BC054736.1 | - | 8 | 1961 | |||
BC071191.1 | - | 7 | 1708 | 6467750 |