Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serinc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253386.1 | NP_001240315.1 | 10 | 1694 | ||
NM_172702.3 | NP_766290.2 | 10 | 1689 | |||
GenBank mRNA | AK075863.1 | - | 10 | 1656 | ||
AK158914.1 | - | 10 | 1695 | |||
BC031720.1 | - | 10 | 1670 | 4209575 |