Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lin54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115010.1 | NP_001108482.1 | 6, 7 | 1541 | ||
NM_172714.4 | NP_766302.2 | 6, 7 | 1409 | |||
GenBank mRNA | AK029288.1 | - | 7, 8 | 855 | ||
AK030894.1 | - | 6, 7 | 1543 | |||
AK134320.1 | - | 6, 7 | 1407 | |||
AK220225.1 | - | 6, 7 | 1404 | |||
BC106126.1 | - | 5, 6 | 1045 | |||
BC125580.1 | - | 4, 5 | 536 | |||
BC145025.1 | - | 4, 5 | 536 | 9053573 |