Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcgf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172716.4 | NP_766304.2 | 5 | 485 | ||
GenBank mRNA | AK033609.1 | - | 5 | 560 | ||
AK085574.1 | - | 5 | 541 | |||
AK089091.1 | - | 5 | 486 | |||
BC049266.1 | - | 352 | 5371040 |