Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276287.1 | NP_001263216.1 | 3 | 338 | ||
NM_181066.3 | NP_851411.1 | 3 | 240 | |||
GenBank mRNA | AK043212.1 | - | 3 | 203 | ||
AK049321.1 | - | 3 | 229 | |||
AK084987.1 | - | 3 | 229 | |||
AK085091.1 | - | 3 | 229 | |||
AK142387.1 | - | 3 | 200 | |||
AK154448.1 | - | 3 | 211 | |||
AK170269.1 | - | 3 | 232 | |||
AK187162.1 | - | 2 | 130 | |||
BC068301.1 | - | 3 | 304 | 30061848 | ||
BY788789.1 | - | 3 | 339 |