Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E130309D02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172726.4 | NP_766314.3 | 1 | 421 | ||
GenBank mRNA | AK041667.1 | - | 1 | 367 | ||
AK050733.1 | - | 1 | 335 | |||
AK053801.1 | - | 1 | 422 | |||
AK054218.1 | - | 1 | 122 | |||
AK086571.1 | - | 1 | 404 | |||
AK087531.1 | - | 1 | 422 | |||
AK144964.1 | - | 1 | 402 | |||
AK172614.1 | - | 1 | 368 | |||
BC058940.1 | - | 1 | 199 | |||
BC079853.1 | - | 1 | 355 | 5716912 | ||
BC082321.1 | - | 1 | 165 | 6411746 |