Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3hc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172735.2 | NP_766323.1 | 1 | 131 | ||
GenBank mRNA | AJ537495.1 | - | 1 | 123 | ||
AK088527.1 | - | 1 | 132 | |||
AK146228.1 | - | 1 | 146 | |||
AK151694.1 | - | 1 | 127 | |||
AK164393.1 | - | 1 | 130 | |||
AK167465.1 | - | 1 | 132 | |||
AK167992.1 | - | 1 | 146 | |||
AK168071.1 | - | 1 | 142 | |||
BC037445.1 | - | 1 | 85 | |||
BC050141.1 | - | 1 | 124 | 30011212 |