Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Heatr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172757.3 | NP_766345.3 | 4 | 581 | ||
GenBank mRNA | AK047782.1 | - | 4 | 612 | ||
AK052635.1 | - | 4 | 608 | |||
AK134510.1 | - | 4 | 585 | |||
AK136424.1 | - | 4 | 591 | |||
AK140522.1 | - | 4 | 599 | |||
AK163715.1 | - | 4 | 568 | |||
AK165541.1 | - | 4 | 476 | |||
BC065060.1 | - | 4 | 581 | 6402217 |