Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172758.4 | NP_766346.1 | 4, 5 | 714 | ||
GenBank mRNA | AK052548.1 | - | 4, 5 | 715 | ||
AK171137.1 | - | 4, 5 | 722 | |||
AK218537.1 | - | 1, 2 | 64 |