Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276452.1 | NP_001263381.1 | 2 | 329 | ||
NM_172773.3 | NP_766361.1 | 2 | 329 | |||
GenBank mRNA | AK029102.1 | - | 2 | 312 | ||
AK087395.1 | - | 2 | 312 | |||
AK169868.1 | - | 2 | 278 | |||
BC058785.1 | - | 2 | 299 | 6309785 |