Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Maob | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172778.2 | NP_766366.2 | 2 | 220 | ||
GenBank mRNA | AK031833.1 | - | 2 | 204 | ||
AK054050.1 | - | 2 | 219 | |||
BC113182.1 | - | 2 | 209 | 40061991 | ||
BC113788.1 | - | 2 | 209 | 40061990 | ||
BY257414.1 | - | 2 | 221 |