Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172780.3 | NP_766368.2 | 2 | 505 | ||
GenBank mRNA | AK032326.1 | - | 2 | 486 | ||
AK122232.1 | - | 2 | 476 | |||
BC130221.1 | - | 2 | 490 | 40141414 |