Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nxt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161430.2 | NP_001154902.1 | 4 | 274 | ||
NM_001290532.1 | NP_001277461.1 | 4 | 359 | |||
NM_001290533.1 | NP_001277462.1 | 3 | 365 | |||
NM_172782.4 | NP_766370.2 | 3 | 311 | |||
GenBank mRNA | AK032102.1 | - | 3 | 282 | ||
AK144349.1 | - | 3 | 295 | |||
BB851047.1 | - | 3 | 313 | |||
BB857403.1 | - | 4 | 275 | |||
BC068166.1 | - | 3 | 264 | 30361776 | ||
BC094570.1 | - | 3 | 228 | 30361291 | ||
BC118503.1 | - | 3 | 239 | 40106658 | ||
CA482101.1 | - | 4 | 310 | |||
CJ148018.1 | - | 4 | 360 |