Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sarm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168521.1 | NP_001161993.1 | 7 | 2236 | ||
NM_172795.3 | NP_766383.2 | 7 | 2116 | |||
GenBank mRNA | AK044113.1 | - | 2 | 453 | ||
AK172964.1 | - | 6 | 2379 | |||
AY444167.1 | - | 7 | 1940 | |||
BC052098.1 | - | 7 | 1762 | |||
BC058534.1 | - | 7 | 2063 | 6834056 | ||
BC080850.1 | - | 7 | 2116 | 6849138 | ||
BU704378.1 | - | 3 | 487 |